Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYOM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120745
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120745 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120745 Supplier Invitrogen™ Supplier No. PA5120745
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse heart, Mouse skeletal muscle, Rat heart, Rat skeletal muscle. Immunogen sequence: MSLVTVPFYQ KRHRHFDQSY RNIQTRYLLD EYASKKRAST QASSQKSLSQ RSSSQRASSQ TSLGGTICRV CAKRVSTQED EEQENRSRYQ SLVAAYGEAK RQRFLSELAH LEEDVHLARS QARDKLDKYA IQQMMEDKLA WERHTFEERI SRAPEILVRL RSHTVWERMS VKLCFTVQGF PTPVVQWYKD GSLICQAAEP.

The giant protein titin, together with its associated proteins, interconnects the major structure of sarcomeres, the M bands and Z discs. The C-terminal end of the titin string extends into the M line, where it binds tightly to M-band constituents of apparent molecular masses of 190 kD and 165 kD. The predicted MYOM2 protein contains 1,465 amino acids. Like MYOM1, MYOM2 has a unique N-terminal domain followed by 12 repeat domains with strong homology to either fibronectin type III or immunoglobulin C2 domains. Protein sequence comparisons suggested that the MYOM2 protein and bovine M protein are identical. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYOM2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MYOM2
Gene Accession No. P54296
Gene Alias 165 kDa connectin-associated protein; 165 kDa titin-associated protein; AW146149; M-band protein; M-protein; MYOM2; myomesin (M-protein) 2, 165kDa; myomesin 2; myomesin family member 2; Myomesin-2; skeletal muscle 165kD protein; skeletal muscle 165kDa protein; titin-associated protein, 165 kD; TTNAP
Gene Symbols MYOM2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17930, 306616, 9172
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.