Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYOM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555774 Supplier Invitrogen™ Supplier No. PA555774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASLKVSCTYK EDEGLYMVRV PSPFGPREQS TYVLVRDAEA ENPGAPGSPL NVRCLDVNRD CLILTWAPPS DTRGNPITAY TIERCQGESG EWI Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

MYOM3 (myomesin 3) is a structural component of the M-band in striated muscle and is involved in sarcomere stability and resistance during intense or sustained stretching. MYOM3 can be detected mainly in intermediate speed fibers of skeletal muscle. Recently high level of MYOM3 fragments were detected in sera from patients with muscular dystrophy, including Duchenne muscular dystrophy (DMD). MYOM3 fragments may be used as serum biomarker for DMD and other neuromuscular disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYOM3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Myom3
Gene Accession No. Q5VTT5
Gene Alias 8430427K15Rik; Gm434; MYOM3; myomesin 3; Myomesin family member 3; myomesin family, member 3; myomesin-3
Gene Symbols Myom3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYOM3. Recombinant protein control fragment (Product #RP-94256).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127294
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.