Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYOM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139785
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: EDDSILDLTG DALDAIFTEL GRIGALSATP LKIQGTEEGI RIFSKVKYYN VEYMKTTWFH KDKRLESGDR IRTGTTLDEI WLHILDPKD.
MYOM3 (myomesin 3) is a structural component of the M-band in striated muscle and is involved in sarcomere stability and resistance during intense or sustained stretching. MYOM3 can be detected mainly in intermediate speed fibers of skeletal muscle. Recently high level of MYOM3 fragments were detected in sera from patients with muscular dystrophy, including Duchenne muscular dystrophy (DMD). MYOM3 fragments may be used as serum biomarker for DMD and other neuromuscular disorders.
Specifications
| MYOM3 | |
| Polyclonal | |
| Unconjugated | |
| Myom3 | |
| 8430427K15Rik; Gm434; MYOM3; myomesin 3; Myomesin family member 3; myomesin family, member 3; myomesin-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 127294 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VTT5 | |
| Myom3 | |
| Recombinant protein corresponding to Human MYOM3. Recombinant protein control fragment (Product #RP-108780). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction