Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myomegalin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119704
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119704 Supplier Invitrogen™ Supplier No. PA5119704
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: NIH/3T3 cell lysates, rat skeletal muscle tissue, mouse heart tissue. Subcellular Location: Cytoplasm, Cytoskeleton, Golgi apparatus.

The protein encoded by this gene serves to anchor phosphodiesterase 4D to the Golgi/centrosome region of the cell. Defects in this gene may be a cause of myeloproliferative disorder (MBD) associated with eosinophilia. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myomegalin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 50% glycerol, 1% BSA and 0.05% sodium azide; pH 7.4
Gene PDE4DIP
Gene Accession No. Q80YT7, Q9WUJ3
Gene Alias 4732458A06Rik; 9430063L05Rik; C87016; cardiomyopathy-associated protein 2; CMYA2; D130016K21Rik; D3Bwg1078e; KIAA0454; KIAA0477; mKIAA0454; MMGL; Myomegalin; myomegalin/phosphodiesterase 4D interacting protein variant 8; Pbp46; PDE4DIP; phosphodiesterase 4D interacting protein; phosphodiesterase 4D interacting protein (myomegalin); phosphodiesterase 4D-interacting protein; Phosphodiesterase-binding protein clone 46; upregulated during skeletal muscle growth 4; Usmg4
Gene Symbols PDE4DIP
Host Species Rabbit
Immunogen Synthetic peptide within rat Myomegalin aa 2130-2190.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64183, 83679
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.