Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myoneurin Antibody (OTI3B9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272864F
Description
Myoneurin Monoclonal specifically detects Myoneurin in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Myoneurin | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
MYNN | |
Recombinant protein fragment corresponding to amino acids 10-300 of human Myoneurin (NP_061127) produced in E.coli. | |
0.1 mL | |
55892.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI3B9 | |
Flow Cytometry | |
myoneurin, OSZF, ZBTB31SBBIZ1, Zinc finger and BTB domain-containing protein 31, zinc finger protein with BTB/POZ domain, ZNF902 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction