Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myoregulin Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc MYREGLN101AP

Description
Myoregulin Polyclonal antibody specifically detects Myoregulin in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
Myoregulin | |
Polyclonal | |
Unconjugated | |
MRLN | |
MLN; MRLN; Myoregulin | |
Synthetic peptide taken within amino acid region 1-50 on human Myoregulin protein. | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
P0DMT0, Q9CV60 | |
Rabbit | |
Affinity chromatography | |
RUO | |
100507027, 100911740, 69563 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction