Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin 1A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Myosin 1A |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Myosin 1A Polyclonal specifically detects Myosin 1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Myosin 1A | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
BBMI, BBM-I, Brush border myosin I, brush border myosin-I, DFNA48, MIHC, MYHL, Myosin I heavy chain, myosin IA, myosin, heavy polypeptide-like (100kD), myosin-Ia | |
MYO1A | |
IgG | |
Affinity Purified |
Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Vision | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
4640 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FSLHLSEMSSVGSKGDFLLVSEHVIELLTKMYRAVLDATQRQLTVTVTEKFSVRFKENSVAVKVVQGPAGGDNSKLRY | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title