Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin 1GRabbit anti-Chimpanzee, Human, Mouse, Rat, Polyclonal, Rockland™
SDP

Catalog No. 501991884
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1884 Supplier Rockland Immunochemicals Supplier No. 600401959
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity-purified antibody is directed against human Myosin 1G protein. A BLAST analysis was used to suggest cross reactivity with Myosin 1G from human, mouse, chimpanzee and rat based on 100% homology with the immunizing sequence. Cross reactivity with Myosin 1G from other sources has not been determined.

Specifications

Antigen Myosin 1G
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene MYO1G
Gene Accession No. B0I1T2, Q5SUA5
Gene Alias HA2; HLA-HA2; MHAG; mHag HA-2; Minor histocompatibility antigen HA-2; MYO1G; myosin IG; myosin-Ig; Unconventional myosin-Ig
Gene Symbols MYO1G
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246177, 289785, 463387, 64005
Target Species Chimpanzee, Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.