Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Myosin-3 (MYH3), Mouse, Unlabeled, Clone: BF-G6,

Catalog No. MABT818MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABT818MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT818MI Supplier MilliporeSigma™ Supplier No. MABT818
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myosin-3 (UniProt A6QPA6; also known as Muscle embryonic myosin heavy chain, Myosin heavy chain 3, MYH3, Protein, Myosin heavy chain, fast skeletal muscle, embryonic, SMHCE) is encoded by the MYH3 gene (Gene ID 281338) in bovine species. Myosin is a major contractile protein that converts chemical energy into mechanical energy through the hydrolysis of ATP. MYH3 is predominantly expressed during fetal development and its expression declines rapidly after birth. It is not normally present in adult muscle except during muscle regeneration. Mutations in MYH3 are known cause two congenital contracture syndromes, Freeman-Sheldon syndrome and Sheldon-Hall syndrome, the most severe forms of distal arthrogryposis (DA). Ref.: Pokrzywa, M., et al. (2015). PLoS One. 10(11):e0142094; Toydamir, R.M., et al. (2006). Nature Genet. 38(5):561-565.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin-3 (MYH3)
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone BF-G6
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene MYH3
Gene Accession No. A6QPA6
Host Species Mouse
Immunogen Fetal bovine skeletal muscle myosin (Schiaffino, S., et al. (1986). Exp. Cell Res. 163(1):211-220).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001095305
Test Specificity Clone BF-G6 detected myosin in human fetal muscle, but not adult human skeletal muscle tissue (Schiaffino, S., et al. (1986). Exp. Cell Res. 163(1):211-220).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.