Learn More
Invitrogen™ Myosin 4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143721
Description
The antibody detects a 215 kDa* protein corresponding to the apparent molecular mass of MYH4 on SDS-PAGE immunoblots of mouse extraocular muscle and mouse C2C12 cells. The antibody may also detect other skeletal MYHs, such as MYH1 and MYH2, which have significant homology to the C-terminal region of MYH4.
Target protein includes a fast myosin isoform expressed in skeletal muscle and has function involved in Muscle contraction.
Specifications
Myosin 4 | |
Polyclonal | |
Unconjugated | |
MYH4 | |
2B myosin heavy chain; AI506973; fast myosin heavy chain 2B; LOW QUALITY PROTEIN: myosin-4; MHC; MHC2B; Minimsc; Minmus; MM; MYH1; MYH2B; MYH-2B; myh4; MyHC IIb isoform; myHC-2b; MyHC-IIb; Myhsf; myosin heavy chain; myosin heavy chain 1; myosin heavy chain 2b; myosin heavy chain 2X; myosin heavy chain 4; Myosin heavy chain IIb; myosin heavy chain IIb isoform; myosin heavy chain isoform 2b; myosin heavy chain type IIb; Myosin heavy chain, skeletal muscle; myosin heavy chain, skeletal muscle, fetal; Myosin heavy chain, skeletal muscle, juvenile; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 4, skeletal muscle; myosin-4; unnamed protein product | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17884, 360543, 4622 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.125 mg/mL | |
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
Q29RW1, Q5SX39, Q9Y623 | |
MYH4 | |
A synthetic peptide (coupled to KLH) corresponding to amino acid residues at the C-terminus from mouse MYH4/MyHC-IIB. This sequence has 100% identity with rat and human MYH4, and has significant homology to MYH1, MYH2, and MYH8. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.