Learn More
Myosin-6 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | Myosin-6 |
---|---|
Concentration | 25 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0132
![]() ![]() |
Rockland Immunochemicals
100401J03S |
25 μL |
Each for $174.73
|
|
|||||
50-199-0131
![]() ![]() |
Rockland Immunochemicals
100401J03 |
100 μL |
Each for $636.24
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-Myosin-6 is directed against the myosin-6 protein. A BLAST analysis was used to suggest reactivity with 94% human Myosin VI. Cross-reactivity with myosin6 from other sources have not been determined.
This gene encodes a protein involved intracellular vesicle and organelle transport, especially in the hair cell of the inner ear. Mutations in this gene have been found in patients with non-syndromic autosomal dominant and recessive hearing loss.Specifications
Myosin-6 | |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Pig | |
Q29122, Q9UM54 | |
397085, 4646 | |
Anti-Myosin-6 was prepared from whole rabbit serum produced by repeated immunizations with full length myosin-6 construct expressed in SF9 insect cells corresponding to porcine myosyn-6 protein. | |
-20°C, Avoid Freeze/Thaw Cycles | |
MYO6 |
25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
BC029719; DFNA22; DFNB37; KIAA0389; MYO6; myosin VI; myosin-VI; RGD1560646; Snell's waltzer; sv; tailchaser; Tlc; Unconventional myosin-6; Unconventional myosin-VI | |
MYO6 | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.