Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin 7 (MYH7) Mouse anti-Human, Mouse, Clone: BA-D5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT838100UG
Description
Anti-Myosin 7 (MYH7), clone BA-D5 is a mouse monoclonal antibody that detects Myosin-7 and has been tested for use in Immunofluorescence, Immunohistochemistry, and Western Blotting.
Specifications
Myosin 7 (MYH7) | |
Monoclonal | |
Please refer to lot specific datasheet. | |
Myh7 | |
Purified myosin from bovine atrium. | |
100 μg | |
Cell Structure | |
NP_777152 | |
Human, Mouse | |
Purified |
Immunofluorescence, Immunohistochemistry, Western Blot | |
BA-D5 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone BA-D5 detects Myosin-7 in multiple mammalian species. It detects both alpha- and beta-slow myosin heavy chain. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction