Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin 7 (MYH7) Rat anti-Mouse, Clone: 4B51E8, MilliporeSigma™

Catalog No. MABT849100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT849-100 100 μg
MABT849-25UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT849-100 Supplier MilliporeSigma Supplier No. MABT849100UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Myosin 7 (MYH7), clone 4B51E8 is a rat monoclonal antibody that detects Mysoin-7 and has been tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin 7 (MYH7)
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4B51E8
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Myh7
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to 39 amino acids from the C-terminal region of murine Mysoin-7.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone 4B51E8 is a rat monoclonal antibody that detects murine Myosin-7. It targets an epitope within 39 amino acids from the C-terminall half.
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from the date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.