Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin-9 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501567854
Encompass_Preferred
Change view
Click to view available options
20 μL; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-156-7854 20 μL; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7854 Supplier Bethyl Laboratories, Inc Supplier No. A304490AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

This gene encodes a myosin IIA heavy chain that contains an IQ domain and a myosin head-like domain. The protein is involved in several important functions, including cytokinesis, cell motility and maintece of cell shape. Defects in MYH9 are the cause of non-syndromic sensorineural deafness autosomal domit type 17, Epstein syndrome, Alport syndrome with macrothrombocytopenia, Sebastian syndrome, Fechtner syndrome and macrothrombocytopenia with progressive sensorineural deafness.

Specifications

Antigen Myosin-9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MYH9
Gene Accession No. P35579, Q8VDD5
Gene Alias BDPLT6, cellular myosin heavy chain, type A, DFNA17, EPSTS, FTNS, MHA, Myosin heavy chain 9, Myosin heavy chain, non-muscle IIa, myosin, heavy polypeptide 9, non-muscle, NMHC-II-A, NMMHC II-a, NMMHCA, NMMHC-A, NMMHC-IIA, non-muscle myosin heavy chain 9, non-muscle myosin heavy chain A, non-muscle myosin heavy chain IIa, nonmuscle myosin heavy chain II-A, non-muscle myosin heavy polypeptide 9
Gene Symbols MYH9
Host Species Rabbit
Immunogen Between 1900 and 1950
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17886, 4627
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.