Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myosin heavy chain 1 Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5145318
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145318 20 μL
FEPA5145317 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145318 Supplier Invitrogen™ Supplier No. PA5145318
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Myosin is a protein which is known to interact with actin in muscle and non-muscle cells. It contains two identical heavy chains and four light chains. Myosin molecules consist of two major regions: tails (rods) and heads. They aggregate into filaments through the tail region and interact with actin and with ATP through the head region. Multiple forms of myosin heavy chains exist for each muscle type-skeletal, cardiac, smooth and non-muscle isomyosin forms exist in different types of skeletal muscle, depending on the physiological function of the muscle. They are designated at type I (slow twitch) and type II (fast twitch).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin heavy chain 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Myh1
Gene Accession No. P12882
Gene Alias A530084A17Rik; epididymis luminal protein 71; HEL71; IId; IId/x; MdMs; MHC2X/D; MHC-2X/D; MYH1; MYHa; MYHC; myHC-2x; MyHC-2X/D; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; MYHSA1; Myhs-f; Myhsf2; Myhs-f2; myosin heavy chain 1; Myosin heavy chain 2x; myosin heavy chain IIx/d; Myosin heavy chain, skeletal muscle, adult 1; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin-1; type 2X myosin heavy chain
Gene Symbols Myh1
Host Species Rabbit
Immunogen Recombinant Human MYH1 protein, Tyr585-Ala794 (Accession #P12882).
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4619
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.