Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin heavy chain 11 Antibody (160), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289243AF647
Description
Myosin heavy chain 11 Monoclonal specifically detects Myosin heavy chain 11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Myosin heavy chain 11 | |
Monoclonal | |
Alexa Fluor 647 | |
AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain | |
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Myosin heavy chain 11. | |
0.1 mL | |
Hypoxia, Stem Cell Markers | |
4629 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
160 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction