Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin heavy chain 11 Rabbit anti-Human, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Rabbit, Clone: MYH11/2303R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$304.50

Specifications

Antigen Myosin heavy chain 11
Clone MYH11/2303R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122664
SDP
View Documents
Novus Biologicals
NBP30759520UG
20 μg
Each of 1 for $304.50
Only null left
Add to Cart
 
Description

Description

Myosin heavy chain 11 Monoclonal specifically detects Myosin heavy chain 11 in Human, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Rabbit samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Myosin heavy chain 11
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Hypoxia, Stem Cell Markers
10 mM PBS with 0.05% BSA
4629
IgG
Protein A or G purified
MYH11/2303R
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human, Rat, Pig, Bovine, Canine, Chicken, Feline, Guinea Pig, Rabbit
AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain
Recombinant full-length human Myosin heavy chain 11 protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.