Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin heavy chain 2 Antibody (TH81 (BGN/04/4481)), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065675UV
Description
Myosin heavy chain 2 Monoclonal specifically detects Myosin heavy chain 2 in Human, Rat, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Myosin heavy chain 2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MYH2 | |
Myosin heavy chain from rabbit muscle | |
0.1 mL | |
Cytoskeleton Markers | |
4620 | |
Human, Rat, Rabbit | |
Purified |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
TH81 (BGN/04/4481) | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
fast 2a myosin heavy chain, MYH2A, MYHas8, MyHC-2A, MyHC-IIa, MYHSA2, myosin heavy chain 2, myosin heavy chain 2a, myosin heavy chain IIa, myosin, heavy chain 2, skeletal muscle, adult, myosin, heavy polypeptide 2, skeletal muscle, adult, myosin-2, skeletal muscle, adult 2, type IIA myosin heavy chain | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
NB100-65675 reacts strongly with human cardiac myosin and also recognizes rabbit myosin heavy and light chains. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction