Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myosin Heavy Chain Recombinant Rabbit Monoclonal Antibody (6V0G1)

Rabbit Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA535613

Catalog No. PIMA535613


Only null left
Add to Cart

Description

Description

Immunogen sequence: VAELGEQIDN LQRVKQKLEK EKSEFKLELD DVTSNMEQII KAKANLEKMC RTLEDQMNEH RSKAEETQRS VNDLTSQRAK LQTENGELSR QLDEKEALIS QLTRGKLTYT QQLEDLKRQL EEEVKAKNAL AHALQSARHD CDLLREQYEE ETEAKAELQR VLSKANSEVA QWRTKYETDA IQRTEELEEA KKKLAQRLQE A.

The function of Mhc is muscle contraction.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Myosin Heavy Chain
Recombinant Monoclonal
0.4 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P02563, P12882, P13533, Q02566, Q5SX40
MYH1, Myh6
Recombinant fusion protein containing a sequence corresponding to amino acids 1200-1400 of human Myosin heavy chain (P12883).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
6V0G1
Unconjugated
MYH1
A530084A17Rik; A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-MHC; ASD3; beta-myosin heavy chain; Bsh; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; CG17927; CG17927-PA; CG17927-PB; CG17927-PC; CG17927-PD; CG17927-PE; CG17927-PF; CG17927-PG; CG17927-PH; CG17927-PI; CG17927-PK; CG17927-PL; CG17927-PM; CG17927-PN; CG17927-PO; CG17927-PP; CG17927-PQ; CG17927-PR; CG17927-PS; CG17927-PT; CG17927-PU; CG17927-PV; chain; chr2L:16765388..16765564; chr2L:16781880..16782040; CMD1EE; CMH14; DFNA4; DFNA4A; Dm II; Dmel\CG17927; Dmel_CG17927; DmMHC; DROMHC; DroMII; epididymis luminal protein 71; FP17425; heavy chain; HEL71; ifm(2)2; ifm(2)RU1; ifm(2)RU2; IId; IId/x; indirect flight muscle (2) RU1; indirect flight muscle (2) RU2; KIAA2034; l(2)36Ae; l(2)k10423; l(2)M66; MdMs; mhc; MHC16; MHC2X/D; MHC-2X/D; Mhc36B; Mhc-PA; Mhc-PB; Mhc-PC; Mhc-PD; Mhc-PE; Mhc-PF; Mhc-PG; Mhc-PH; Mhc-PI; Mhc-PK; Mhc-PL; Mhc-PM; Mhc-PN; Mhc-PO; Mhc-PP; Mhc-PQ; Mhc-PR; Mhc-PS; Mhc-PT; Mhc-PU; Mhc-PV; mMHC; MRP; muscle myosin; muscle myosin heavy chain; muscle myosin II; muscle myosin II heavy chain; muscle myosin-II; muscle specific myosin II; MYH1; MYH14; MYH14 variant protein; MYH17; MYH6; MYHa; MYHC; MyHC-2x; MyHC-2X/D; Myhca; Myhc-a; MyHC-alpha; MyHC-IId/x; MYHC-IIX; myHC-IIx/d; MYHSA1; Myhs-f; Myhsf2; Myhs-f2; Myo; Myo6; myosin; myosin HC; myosin heavy; myosin heavy chain; myosin heavy chain 1; Myosin heavy chain 14; Myosin heavy chain 2x; myosin heavy chain 6; Myosin heavy chain IIx/d; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; Myosin heavy chain, muscle; Myosin heavy chain, non-muscle IIc; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin heavy chain, skeletal muscle, adult 1; myosin heavy Chain-PF; myosin heavy-chain; myosin heavy-chain protein; myosin II heavy chain; myosin rod protein; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 14, non-muscle; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 14; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin-1; Myosin-14; myosin-6; myosin-heavy chain; NMHC II-C; NMHC-II-C; non-muscle myosin heavy chain IIc; nonmuscle myosin heavy chain II-C; Not-upheld; Nup; PNMHH; samba; sarcomeric myosin heavy-chain; shrunken thorax; Shrunken-thorax; Sht; sMHC; SSS3; Stp; stuckup; type 2X myosin heavy chain
Rabbit
Affinity chromatography
RUO
17879, 17888, 287408, 29556, 4619, 4624
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.