Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Heavy Chain Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB4470 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4470 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4470 Supplier R&D Systems Supplier No. MAB4470
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 57 publications

Myosin Heavy Chain Monoclonal specifically detects Myosin Heavy Chain in Chicken∣Goat∣Human∣Mouse∣Multi-species∣Pig∣Rat∣Transgenic Mouse∣Xenograft∣Zebrafish samples. It is validated for Bioassay∣Flow Cytometry∣Immunocytochemistry∣Immunocytochemistry/Immunofluorescence∣Immunofluorescence∣Immunohistochemistry∣Immunohistochemistry (Paraffin)∣Immunoprecipitation∣Neutralization∣Simple Western∣Western Blot.

Specifications

Antigen Myosin Heavy Chain
Applications Bioassay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Simple Western, Western Blot
Classification Monoclonal
Clone MF20
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Gene Alias DFNA4, FLJ13881, FLJ43092, KIAA2034DKFZp667A1311, MHC16, MYH14 variant protein, MYH17, MYHC, myosin, Myosin heavy chain 14, Myosin heavy chain, non-muscle IIc, myosin, heavy chain 14, myosin, heavy chain 14, non-muscle, myosin, heavy polypeptide 14, myosin-14, NMHC II-C, NMHC-II-C, nonmuscle myosin heavy chain II-C, Non-muscle myosin heavy chain IIc
Gene Symbols MYH14
Host Species Mouse
Immunogen Chicken pectoralis-derived Myosin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79784
Test Specificity Detects Myosin Heavy Chain in human, mouse, rat and other mammalian, avian, and amphibian species.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Chicken, Goat, Human, Mouse, Multi-species, Pig, Rat, Transgenic Mouse, Xenograft, Zebrafish
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Form Purified
Isotype IgG2b
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.