Learn More
Myosin Heavy Chain, Cardiac Monoclonal Antibody (9G11), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 603820

Description
Positive control: Cardiac muscle Cellular location: Cytoplasmic.
Myosin Heavy Chain, Cardiac Monoclonal antibody specifically detects Myosin Heavy Chain, Cardiac in Bovine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
Myosin Heavy Chain, Cardiac | |
Monoclonal | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
A2AQP0, A7E2Y1, P02563, P02564, P12883, P13533, Q02566, Q91Z83, Q9BE39 | |
Mouse | |
Protein A | |
RUO | |
100296004, 140781, 17888, 282714, 29556, 29557, 311570, 4624, 4625, 521764, 57644, 668940 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
9G11 | |
Unconjugated | |
Myh7 | |
A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-cardiac myosin heavy chain; alpha-MHC; alpha-myosin; alpha-myosin heavy chain; alpha-myosin heavy chain subfragment 2 exon; antigen MLAA-21; ASD3; beta cardiac myosin heavy chain; beta myosin heavy chain; beta-MHC; beta-myosin; beta-myosin heavy chain; B-MHC; cardiac muscle myosin heavy chain 7 beta; cardiac myosin heavy chain alpha isoform; cardiac myosin heavy chain beta isoform; cardiomyopathy, hypertrophic 1; CMD1EE; CMD1S; CMH1; CMH14; KIAA1512; LOC100736765; LOW QUALITY PROTEIN: myosin-6; LOW QUALITY PROTEIN: myosin-7B; MHC14; MPD1; Myh14; MYH6; MYH7; Myh7b; MYH-beta/slow; MYHC; Myhca; Myhc-a; MyHC-alpha; MYHCB; Myhc-b; MyHC-beta; MyHC-I; MyHC-slow; myopathy, distal 1; myosin alpha heavy chain; Myosin cardiac muscle beta chain; myosin heavy chain; myosin heavy chain 6; Myosin heavy chain 7; myosin heavy chain 7B; myosin heavy chain 7B, cardiac muscle beta isoform; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain polypeptide 7 cardiac muscle fetal; myosin heavy chain slow; myosin heavy chain slow isoform; myosin heavy chain slow type 1 (beta cardiac); myosin heavy chain, cardiac; myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle beta isoform; myosin heavy chain, cardiac muscle, adult; myosin heavy chain, cardiac muscle, fetal; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin heavy chain, polypeptide 7; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy chain 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin, heavy chain 7, cardiac muscle, beta; myosin, heavy chain 7B, cardiac muscle, beta; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin, heavy polypeptide 7, cardiac muscle, beta; myosin, heavy polypeptide 7B, cardiac muscle, beta; myosin-6; Myosin-7; myosin-7B; rhabdomyosarcoma antigen MU-RMS-40.7A; slow A MYH14; slow myosin heavy chain; slow myosin heavy chain-beta; slow type myosin heavy chain; SPMD; SPMM; SSS3; type slow/beta myosin heavy chain; U937-associated antigen; Unknown (protein for IMAGE:7985957) | |
The recombinant protein of human myosin heavy chain recombinant protein | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.