Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myosin Heavy Chain Recombinant Rabbit Monoclonal Antibody (JF097-7)
GREENER_CHOICE

Catalog No. PIMA532555
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532555 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532555 Supplier Invitrogen™ Supplier No. MA532555
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The function of Mhc is muscle contraction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myosin Heavy Chain
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JF097-7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene Myh6
Gene Accession No. P02563, P12882, P13533, Q02566, Q5SX40
Gene Alias A530084A17Rik; A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-MHC; ASD3; beta-myosin heavy chain; Bsh; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; CG17927; CG17927-PA; CG17927-PB; CG17927-PC; CG17927-PD; CG17927-PE; CG17927-PF; CG17927-PG; CG17927-PH; CG17927-PI; CG17927-PK; CG17927-PL; CG17927-PM; CG17927-PN; CG17927-PO; CG17927-PP; CG17927-PQ; CG17927-PR; CG17927-PS; CG17927-PT; CG17927-PU; CG17927-PV; chain; chr2L:16765388..16765564; chr2L:16781880..16782040; CMD1EE; CMH14; DFNA4; DFNA4A; Dm II; Dmel\CG17927; Dmel_CG17927; DmMHC; DROMHC; DroMII; epididymis luminal protein 71; FP17425; heavy chain; HEL71; ifm(2)2; ifm(2)RU1; ifm(2)RU2; IId; IId/x; indirect flight muscle (2) RU1; indirect flight muscle (2) RU2; KIAA2034; l(2)36Ae; l(2)k10423; l(2)M66; MdMs; mhc; MHC16; MHC2X/D; MHC-2X/D; Mhc36B; Mhc-PA; Mhc-PB; Mhc-PC; Mhc-PD; Mhc-PE; Mhc-PF; Mhc-PG; Mhc-PH; Mhc-PI; Mhc-PK; Mhc-PL; Mhc-PM; Mhc-PN; Mhc-PO; Mhc-PP; Mhc-PQ; Mhc-PR; Mhc-PS; Mhc-PT; Mhc-PU; Mhc-PV; mMHC; MRP; muscle myosin; muscle myosin heavy chain; muscle myosin II; muscle myosin II heavy chain; muscle myosin-II; muscle specific myosin II; MYH1; MYH14; MYH14 variant protein; MYH17; MYH6; MYHa; MYHC; MyHC-2x; MyHC-2X/D; Myhca; Myhc-a; MyHC-alpha; MyHC-IId/x; MYHC-IIX; myHC-IIx/d; MYHSA1; Myhs-f; Myhsf2; Myhs-f2; Myo; Myo6; myosin; myosin HC; myosin heavy; myosin heavy chain; myosin heavy chain 1; Myosin heavy chain 14; Myosin heavy chain 2x; myosin heavy chain 6; Myosin heavy chain IIx/d; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; Myosin heavy chain, muscle; Myosin heavy chain, non-muscle IIc; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin heavy chain, skeletal muscle, adult 1; myosin heavy Chain-PF; myosin heavy-chain; myosin heavy-chain protein; myosin II heavy chain; myosin rod protein; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 14, non-muscle; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 14; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin-1; Myosin-14; myosin-6; myosin-heavy chain; NMHC II-C; NMHC-II-C; non-muscle myosin heavy chain IIc; nonmuscle myosin heavy chain II-C; Not-upheld; Nup; PNMHH; samba; sarcomeric myosin heavy-chain; shrunken thorax; Shrunken-thorax; Sht; sMHC; SSS3; Stp; stuckup; type 2X myosin heavy chain
Gene Symbols MYH1, Myh6
Host Species Rabbit
Immunogen Recombinant protein within Human Myosin heavy chain aa 1220-1420.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17879, 17888, 287408, 29556, 4619, 4624
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.