Learn More
Myosin Heavy Chain, (Smooth Muscle) Monoclonal Antibody (ABM201), Abbomax
Mouse Monoclonal Antibody
Supplier: Abbomax Inc 5005244

Description
Positive control: Smooth muscle Cellular location: Cytoplasmic.
Myosin Heavy Chain, (Smooth Muscle) Monoclonal antibody specifically detects Myosin Heavy Chain, (Smooth Muscle) in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Myosin Heavy Chain, (Smooth Muscle) | |
Monoclonal | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P35749 | |
Mouse | |
Protein A | |
RUO | |
4629 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
ABM201 | |
Unconjugated | |
MYH11 | |
AAT4; AV071570; contractile protein; FAA4; KIAA0866; LOW QUALITY PROTEIN: myosin-11; Myh11; myh11 {ECO:0000312; Myosin heavy chain 11; myosin heavy chain 11, smooth muscle; myosin heavy chain 21; myosin heavy chain isoform SMA; myosin heavy chain SMA; myosin heavy chain, smooth muscle isoform; myosin, heavy chain 11, smooth muscle; myosin, heavy polypeptide 11, smooth muscle; myosin-11; RGD:3136}; SM1; SM2; SMHC; SMMHC; smooth muscle myosin heavy chain | |
A synthetic peptide corrsponding to the internal segment of human Myosin | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.