Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Light Chain 2 Antibody (3B2), Biotin, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP130249B

Catalog No. NBP130249B


Only null left
Add to Cart

Description

Description

Myosin Light Chain 2 Monoclonal specifically detects Myosin Light Chain 2 in Human, Mouse, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Myosin Light Chain 2
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
MYL2
Recombinant human MYL2 (1-166aa) purified from E. coli (NP_000423).
0.1 mL
Phospho Specific
4633
Human, Mouse, Xenopus species
Purified
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
3B2
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2
Mouse
Protein G purified
RUO
Primary
This antibody is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human MYL2 protein.
Store at 4C in the dark.
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.