Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin Light Chain 2 Antibody (3B2), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP130249MFV500
Description
Myosin Light Chain 2 Monoclonal antibody specifically detects Myosin Light Chain 2 in Human, Mouse, Xenopus samples. It is validated for Flow CytometrySpecifications
Myosin Light Chain 2 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Xenopus species | |
Purified |
Flow Cytometry | |
3B2 | |
Flow Cytometry | |
cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2 | |
Recombinant human Myosin Light Chain 2 (1-166aa) purified from E. coli (NP_000423). | |
0.1 mL | |
Phospho Specific | |
4633 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction