Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin light chain kinase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP187744
Description
Myosin light chain kinase Polyclonal specifically detects Myosin light chain kinase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Myosin light chain kinase | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
DKFZp686I10125, EC 2.7.11, EC 2.7.11.18, FLJ12216, Kinase-related protein, KRP, light polypeptide kinase, MLCK1, MLCK108, MLCKMLCK210, MSTP083, MYLK1, myosin light chain kinase, smooth muscle, smooth muscle myosin light chain kinase, telokin | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
MYLK | |
This antibody was developed against Recombinant Protein corresponding to amino acids:KCVAKNDAGQAECSCQVTVDDAPASENTKAPEMKSRRPKSSLPPVLGTESDATVKKKPAPKTPPKAAMPPQIIQFPEDQKVRAGESVELFGKVTGTQPITCTWMKFRKQIQESEHMKVENSENGSKLTILAARQEHCGCYTLLVE | |
0.1 mL | |
Protein Kinase | |
4638 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction