Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin Phosphatase 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Myosin Phosphatase 2 |
---|---|
Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000-1:2500 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Myosin Phosphatase 2 Polyclonal specifically detects Myosin Phosphatase 2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Myosin Phosphatase 2 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
4660 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:TQGVTLTDLQEAERTFSRSRAERQAQEQPREKPTDTEGLEGSPEKHEPSAVPATEAGEGQQPWGRSLDEEPICHRLRCPAQPDKPTTPASPSTSRPSLYTSSHLLW | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000-1:2500 | |
Polyclonal | |
Rabbit | |
Human | |
MGC131980, MGC87886, Myosin phosphatase target subunit 2, myosin phosphatase, target subunit 2, Myosin phosphatase-targeting subunit 2, MYPT 2 MYPT2FLJ40942, protein phosphatase 1 regulatory subunit 12B, protein phosphatase 1, regulatory (inhibitor) subunit 12B | |
PPP1R12B | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title