Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Phosphatase, p Thr853 Antibody (SA19), Novus Biologicals™
SDP

Catalog No. NBP229680 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229680 0.1 mL
NB019426 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229680 Supplier Novus Biologicals Supplier No. NBP229680
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Myosin Phosphatase Monoclonal specifically detects Myosin Phosphatase in Human, Chicken samples. It is validated for Western Blot.

Specifications

Antigen Myosin Phosphatase (p Thr853)
Applications Western Blot
Classification Monoclonal
Clone SA19
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation No buffer with No Preservative
Gene Accession No. O14974
Gene Alias MBSmyosin phosphatase, target subunit 1, MGC133042, Myosin phosphatase target subunit 1, Myosin phosphatase-targeting subunit 1, MYPT1protein phosphatase 1 regulatory subunit 12A, protein phosphatase 1, regulatory (inhibitor) subunit 12A, Protein phosphatase myosin-binding subunit
Gene Symbols PPP1R12A
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide containing the sequence.RS[pT]GV., in which [pT] corresponds to phosphorylated threonine at amino acid residue 850 of chicken myosin phosphatase target subunit 1 (MYPT1). Corresponds to Thr853 in human MYPT1.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 4659
Test Specificity MYPT1 phosphorylated on Thr850 doublet at 130kDa weight may be detected.
Target Species Human, Chicken
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.