Learn More
Epredia™ Lab Vision™ Myosin, Skeletal Muscle Ab-2, Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: MYSN02; same as MY-32
Isotype: IgG1
Species Reactivity: Human, Cow, Mouse, Rat, Rabbit, Chicken and Guinea pig.
Epitope: Not determined
Immunogen: Rabbit muscle myosin
Molecular Weight: 480kDa
Positive Control: Skeletal muscle
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Myosin, Skeletal Muscle Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MYSN02; same as MY-32 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Rabbit muscle myosin |
| Quantity | 7 mL |
| Regulatory Status | RUO |
| Research Discipline | Muscle Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.