Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Myosin, Skeletal Muscle Ab-2, Mouse Monoclonal Antibody

Catalog No. MS1236S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1236S1 500 μL Concentrated
MS1236S0 100 μL Concentrated
MS1236S 1 mL Concentrated
MS1236R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1236S1 Supplier Epredia™ Supplier No. MS1236S1
Add to Cart
Add to Cart

Provides accurate, reproducible results with the Epredia™ Myosin, Skeletal Muscle Ab-2, Mouse Monoclonal Antibody.

Myosin is a 480kDa protein known to interact with actin in muscle and in non-muscle cells. It contains two identical heavy chains (200kDa each) and four light chains (15-26kDa). Myosin molecules consist of two major regions: tails (rod) and heads; they aggregate into filaments through the tail region and interact with actin and with ATP through the head region. Multiple forms of myosin heavy chains exist for each muscle type-skeletal, cardiac, smooth and non-muscle isomyosin forms exist in different types of skeletal muscle, depending on the physiological function of the muscle. These are designated at type I (slow-twitch) and type II (fast-twitch).

Host Species: Mouse

Clone: MYSN02; same as MY-32

Isotype: IgG1

Species Reactivity: Human, Cow, Mouse, Rat, Rabbit, Chicken and Guinea pig.

Epitope: Not determined

Immunogen: Rabbit muscle myosin

Molecular Weight: 480kDa

Positive Control: Skeletal muscle

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Myosin, Skeletal Muscle Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MYSN02; same as MY-32
Conjugate Unconjugated
Host Species Mouse
Immunogen Rabbit muscle myosin
Purification Method Skeletal muscle
Quantity 500 μL
Regulatory Status RUO
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Bovine, Chicken, Guinea Pig, Human, Mouse, Rabbit, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.