Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyosinRabbit anti-Human, Mouse, Rat, Polyclonal, Rockland™
SDP

Catalog No. 501991880
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1880 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1880 Supplier Rockland Immunochemicals Supplier No. 600401938
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against the regulatory light chain of smooth and non-muscle myosin. The antibody detects both unphosphorylated and monophosphorylated forms of the protein. Cross reactivity is expected with myosin light chain from human, mouse and rat sources. Reactivity with the protein from other species has not been determined; however, the sequence of the immunogen is nearly identical in mammalian and avian species. BLAST search analysis was used to determine that the smooth and non-muscle forms of myosin regulatory light chain have identical sequences. Cross reactivity is expected.

Myosin regulatory light polypeptide 9 is a highly conserved, ubiquitously expressed protein that in humans is encoded by the MYL9 gene. Myosin, a structural component of muscle, are hexameric proteins consisting of two heavy chains and four light chains, including a single pair of nonphosphorylatable light chain subunits and another pair of phosphorylatable light chain subunits.The protein encoded by this gene is a myosin light chain that may regulate muscle contraction by modulating the ATPase activity of myosin heads. Myosin light chain (Myl) 9 and Myl12 are new functional ligands for CD69. Blockade of CD69-Myl9/12 interactio ameliorates allergic airway inflammation in ovalbumin-induced and house dust mite-induced mouse models of asthma.

Specifications

Antigen Myosin
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene MYL12A
Gene Accession No. O14950, P13832, P18666, P19105, P24844, Q3THE2, Q64122, Q9CQ19
Gene Alias 1500001M02Rik; 20 kDa myosin light chain; 2900073G15Rik; AI327049; AI838292; brain specific myosin regulatory light chain; C77744; C80200; Epididymis secretory protein Li 24; HEL-S-24; LC20; MLC2; MLC-2; MLC20; MLC-2A; MLC-2B; MLC-2C; MLCB; MLC-B; MRCL3; MRLC1; MRLC2; MRLC3; Mrlcb; MYL12A; Myl12b; MYL2B; MYL9; Mylc2b; Mylc2c; myosin light chain 12A; myosin light chain 12B; myosin light chain 9; myosin light chain, regulatory B; myosin light chain, regulatory B-like; myosin light chain, regulatory C; myosin regulatory light chain 1; Myosin regulatory light chain 12A; Myosin regulatory light chain 12B; myosin regulatory light chain 2; Myosin regulatory light chain 2, nonsarcomeric; Myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 20 kDa; Myosin regulatory light chain 2-A, smooth muscle isoform; Myosin regulatory light chain 2-B, smooth muscle isoform; myosin regulatory light chain 3; Myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; Myosin regulatory light chain MRLC2; Myosin regulatory light chain MRLC3; Myosin regulatory light chain RLC-A; myosin regulatory light chain-B; Myosin regulatory light polypeptide 9; Myosin RLC; Myosin RLC-A; Myosin RLC-B; myosin, light chain 12A, regulatory, non-sarcomeric; myosin, light chain 12B, regulatory; myosin, light chain 9, regulatory; myosin, light polypeptide 9, regulatory; myosin, light polypeptide, regulatory, non-sarcomeric (20kD); Myrl2; NMDA receptor-interacting protein; RGD1309537; RLC; Rlc-a; RLC-B; RLC-C; SHUJUN-1
Gene Symbols MYL12A, MYL12B, MYL9
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103910, 10398, 10627, 296313, 501203, 50685, 67268, 67938, 98932
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.