Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myostatin Monoclonal Antibody (6E4B2)
GREENER_CHOICE

Catalog No. PIMA531804
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531804 Supplier Invitrogen™ Supplier No. MA531804
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31804 has been tested in indirect ELISA.

The protein encoded by this gene is a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This gene is thought to encode a secreted protein which negatively regulates skeletal muscle growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myostatin
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6E4B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Mstn
Gene Accession No. O14793
Gene Alias Cmpt; compact; Gdf8; GDF-8; growth differentiation factor 8; growth/differentiation factor 8; H-Myo-Propeptide; H-Myostatin; MSLHP; MSTN; Myostatin
Gene Symbols Mstn
Host Species Mouse
Immunogen Purified recombinant fragment of human MSTN (AA:24-266) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2660
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.