Learn More
Invitrogen™ Myostatin Monoclonal Antibody (6H12)

Description
MA5-15486 targets Myostatin in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15486 immunogen is purified recombinant fragment of Myostatin expressed in E. Coli. . MA5-15486 detects Myostatin which has a predicted molecular weight of approximately 42.7kDa.
Specifications
Specifications
| Antigen | Myostatin |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 6H12 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | Mstn |
| Gene Accession No. | O14793 |
| Gene Alias | Cmpt; compact; Gdf8; GDF-8; growth differentiation factor 8; growth/differentiation factor 8; H-Myo-Propeptide; H-Myostatin; MSLHP; MSTN; Myostatin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.