Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Myostatin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121874
This item is not returnable.
View return policy
Description
Myostatin Polyclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene is a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This gene is thought to encode a secreted protein which negatively regulates skeletal muscle growth.
Specifications
Myostatin | |
Polyclonal | |
Unconjugated | |
Mstn | |
Cmpt; compact; Gdf8; GDF-8; growth differentiation factor 8; growth/differentiation factor 8; H-Myo-Propeptide; H-Myostatin; MSLHP; MSTN; Myostatin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29152 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
O35312 | |
Mstn | |
Recombinant protein Myostatin. The antigen corresponds to amino acid range 267-376 of the target protein. | |
20 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction