Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Myostatin Recombinant Rabbit Monoclonal Antibody (3W2K7)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551309
Description
Myostatin Recombinant Monoclonal Antibody for Western Blot, ELISA
The protein encoded by this gene is a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This gene is thought to encode a secreted protein which negatively regulates skeletal muscle growth.
Specifications
Myostatin | |
Recombinant Monoclonal | |
0.24 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
O08689, O14793, O35312 | |
Mstn | |
Recombinant protein of human GDF-8/Myostatin. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
Liquid |
ELISA, Western Blot | |
3W2K7 | |
Unconjugated | |
Mstn | |
Cmpt; compact; Gdf8; GDF-8; growth differentiation factor 8; growth/differentiation factor 8; H-Myo-Propeptide; H-Myostatin; MSLHP; MSTN; Myostatin | |
Rabbit | |
Affinity chromatography | |
RUO | |
17700, 2660, 29152 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction