Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myotilin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558081 Supplier Invitrogen™ Supplier No. PA558081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSSAFPASP QQHAGSNPGQ RVTTTYNQSP ASFLSSILPS QPDYNSSKIP SAMDSNYQQS SAGQPINAKP SQTANAKPIP RTPDHEIQG Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.

This gene encodes a cystoskeletal protein which plays a significant role in the stability of thin filaments during muscle contraction. This protein binds F-actin, crosslinks actin filaments, and prevents latrunculin A-induced filament disassembly. Mutations in this gene have been associated with limb-girdle muscular dystrophy and myofibrillar myopathies. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myotilin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYOT
Gene Accession No. Q9UBF9
Gene Alias 5530402I04Rik; 57 kDa cytoskeletal protein; LGMD1; LGMD1A; MFM3; Myo; Myofibrillar titin-like Ig domains protein; MYOT; Myotilin; titan-like protein; titin immunoglobulin domain protein; titin immunoglobulin domain protein (myotilin); Ttid; TTOD
Gene Symbols MYOT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Myotilin. Recombinant protein control fragment (Product #RP-96838).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9499
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.