Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myotrophin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554070
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554070 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554070 Supplier Invitrogen™ Supplier No. PA554070
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NGDLDEVKDY VAKGEDVNRT LEGGRKPLHY AADCGQLEIL EFLLLKGADI NAPDKHHITP LLSAVYEGHV SCVKLLLSKG ADKTVKGPDG LTAFEATDNQ AIKALLQ Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

The transcript produced from this gene is bi-cistronic and can encode both myotrophin and leucine zipper protein 6. The myotrophin protein is associated with cardiac hypertrophy, where it is involved in the conversion of NFkappa B p50-p65 heterodimers to p50-p50 and p65-p65 homodimers. This protein also has a potential function in cerebellar morphogenesis, and it may be involved in the differentiation of cerebellar neurons, particularly of granule cells. A cryptic ORF at the 3' end of this transcript uses a novel internal ribosome entry site and a non-AUG translation initiation codon to produce leucine zipper protein 6, a 6. 4 kDa tumor antigen that is associated with myeloproliferative disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myotrophin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MTPN
Gene Accession No. P58546, P62774, P62775
Gene Alias 5033418D15Rik; GCDP; Granule cell differentiation protein; growth factor; Mtpn; Myotrophin; protein V-1; stimulates myocyte growth; V1; V-1; V-1 protein
Gene Symbols MTPN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Myotrophin. Recombinant protein control fragment (Product #RP-92642).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136319, 14489, 79215
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.