Learn More
MYOZ1 Mouse anti-Human, Clone: OTI8A1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808907

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Myozenins may serve as intracellular binding proteins involved in linking Z-disk proteins such as alpha-actinin, gamma-filamin, TCAP/telethonin, LDB3/ZASP and localizing calcineurin signaling to the sarcomere. This protein plays an important role in the modulation of calcineurin signaling, may play a role in myofibrillogenesis.Specifications
MYOZ1 | |
Monoclonal | |
Unconjugated | |
MYOZ1 | |
CS-2, FATZ, MYOZ | |
Mouse | |
Affinity Chromatography | |
RUO | |
58529 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8A1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NP98 | |
MYOZ1 | |
Human recombit protein fragment corresponding to amino acids 106-299 of human MYOZ1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.