Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYOZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100629
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100629 Supplier Invitrogen™ Supplier No. PA5100629
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MYOZ1.

Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYOZ1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MYOZ1
Gene Accession No. Q9JK37, Q9NP98
Gene Alias 2310001N11Rik; alpha-actinin binding protein; AV090278; calcineurin-2; calsarcin 2; calsarcin-2; CS-2; FATZ; filamin-, actinin- and telethonin-binding protein; MYOZ; myoz1; myozenin 1; myozenin precursor; myozenin-1; Protein FATZ; RGD1561064; skeletal muscle-specific protein
Gene Symbols MYOZ1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MYOZ1(Accession Q9NP98), corresponding to amino acid residues T239-G289.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498440, 58529, 59011
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.