Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYOZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543169 Supplier Invitrogen™ Supplier No. PA543169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IYENHPDVFS DSSMDHFQKF LPTVGGQLGT AGQGFSYSKS NGRGGSQAGG Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Sarcomere assembly is regulated by the muscle protein titin. Titin is a giant elastic protein with kinase activity that extends half the length of a sarcomere. It serves as a scaffold to which myofibrils and other muscle related proteins are attached. This gene encodes a protein found in striated and cardiac muscle that binds to the titin Z1-Z2 domains and is a substrate of titin kinase, interactions thought to be critical to sarcomere assembly. Mutations in this gene are associated with limb-girdle muscular dystrophy type 2G.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYOZ1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MYOZ1
Gene Accession No. Q9NP98
Gene Alias 2310001N11Rik; alpha-actinin binding protein; AV090278; calcineurin-2; calsarcin 2; calsarcin-2; CS-2; FATZ; filamin-, actinin- and telethonin-binding protein; MYOZ; myoz1; myozenin 1; myozenin precursor; myozenin-1; Protein FATZ; RGD1561064; skeletal muscle-specific protein
Gene Symbols MYOZ1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MYOZ1 (aa 71-120).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58529
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.