Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYOZ2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557390 Supplier Invitrogen™ Supplier No. PA557390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEELSHLSNR GARLFKMRQR RSDKYTFENF QYQSRAQINH SIAMQNGKVD GSNLEGGSQQ APLTPPNTPD PR Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 85%.

The calcineurin-binding protein Myozenin 2, also designated calsarcin-1, is a member of the calsarcin protein family. Calcineurin is a calcium- and calmodulin-dependent protein phosphatase that is involved in controlling the slow fiber gene expression in skeletal muscle and hypertrophy of cardiac muscle. The calsarcins are sarcomeric proteins that couple calcineurin and muscle activity. In cardiac and skeletal muscle cells, Myozenin 2 binds calcineurin to α-actinin at the Z-line of the sarcomere. During embryogenesis, Myozenin 1 and 2 are expressed in developing muscle. The Myozenin 2 gene maps to chromosome 4q and is expressed specifically in adult cardiac and slow-twitch skeletal muscle, while Myozenin 1 is only detected in fast skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYOZ2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYOZ2
Gene Accession No. Q9NPC6
Gene Alias 1110012I24Rik; C4orf5; calcineurin-binding protein calsarcin-1; calsarcin-1; CMH16; CS-1; Fatz-2; FATZ-related protein 2; muscle-specific protein; myoz2; myozenin 2; myozenin-2; myozenin-like 2; Myozl2
Gene Symbols MYOZ2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYOZ2. Recombinant protein control fragment (Product #RP-96741).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51778
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.