Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYOZ3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558681 Supplier Invitrogen™ Supplier No. PA558681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CPWQEFVSYR DYQSDGRSHT PSPNDYRNFN KTPVPFGGPL VGGTFPRPGT PFIPEPLSGL ELLRLRPSFN RVAQGWVRNL PESEEL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.

The protein encoded by this gene is specifically expressed in the skeletal muscle, and belongs to the myozenin family. Members of this family function as calcineurin-interacting proteins that help tether calcineurin to the sarcomere of cardiac and skeletal muscle. They play an important role in modulation of calcineurin signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYOZ3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYOZ3
Gene Accession No. Q8TDC0
Gene Alias 4833419K08Rik; calsarcin 3; calsarcin-3; CS3; CS-3; Fatz-3; FATZ-related protein 3; FRP3; Myoz3; myozenin 3; myozenin-3
Gene Symbols MYOZ3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYOZ3. Recombinant protein control fragment (Product #RP-96922).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91977
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.