Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myozenin 1 Antibody (OTI4C2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272865
Description
Myozenin 1 Monoclonal specifically detects Myozenin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Myozenin 1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
calsarcin-2, CS-2, FATZ, Filamin-, actinin- and telethonin-binding protein, MYOZ, myozenin, myozenin 1, myozenin-1, Protein FATZ | |
Mouse | |
31.6 kDa | |
100 μg | |
58529 | |
Human, Mouse, Rat | |
Lyophilized |
Western Blot, Immunohistochemistry | |
OTI4C2 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
MYOZ1 | |
Human recombinant protein fragment corresponding to amino acids 106-299 of human MYOZ1(NP_067068) produced in E.coli. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction