Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ MyPIPETMANTM Select Multichannel
GREENER_CHOICE

Catalog No. FP10015STG Shop All Gilson Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
2 to 20 μL
20 to 200 μL
20 to 300 μL
Number of Channels:
8
12
Model:
P12x10
P12x20
P12x200
P12x300
P8x10
P8x20
P8x200
P8x300
This item is not returnable. View return policy

Catalog No. FP10015STG

Supplier: Gilson™ FP10015ST

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Ergonomic, safe, and reliable, MyPIPETMANTM Select offers volume setting choice and optional handle personalization for optimal use in various protocols.

MyPIPETMANTM Select fully-autoclavable, air-displacement pipettes cover a full volume range, including:

  • Variable volume single channel models covering a volume range from 0.2 μL to 10 mL
  • Variable volume multichannel models covering a volume range from 0.5 μL to 300 μL (8 and 12-channel models)

Set and Secure the Volume your Preferred Way

  • Simply adjust the volume by turning the push button of the pipette with the unique, patented TrilockTM volume-locking system.
    • You can set the volume in your preferred way:
      • Free volume setting position for quick and effortless volume adjustment
      • Light ratchet volume setting for precise volume adjustment
      • Volume locked position to secure pipetting

Ergonomic Design

  • Lightweight and well-balanced, MyPIPETMANTM Select has an elegant design with a textured effect for a better grip.
  • With no compromise on quality and performance, the built-in ejection system with an integrated lever provides reduced tip ejection forces compared to direct ejection systems.
  • This patented ejection system reinforces the pipette's ergonomics for convenient use throughout the day.

Personalize your Pipette for an Optimal Use

  • MyPIPETMANTM Select has optional handle printing on demand. Identify your pipette to avoid any risk of contamination by dedicating it to a specific liquid or workflow step and improve your lab's Lean Management process.

Specifications

Compatible Tips PIPETMAN DIAMOND Tips
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color White
Dispensing Speed Manual
Disposable Reusable
For Use With D-5000 Tip
Grip Type Ergonomic, Textured
Material Polypropylene, PVDF, PBT
Model P8x300
Number of Channels 8
Product Line MyPipetman
Programmability No
Sterility Non-sterile
Type Multichannel Adjustable Volume Pipette
Volume (Metric) 20 to 300 μL
Quantity 1
Width (English) 5.98 in
Color Code Green
Fit for Purpose Certified Quality
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.