Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYPN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563966
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563966 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563966 Supplier Invitrogen™ Supplier No. PA563966
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDLSNNGSLH SANSTTNLAA IEPQPSPPHS EPPSVEQPPK PKLEGVLVNH NEPRSSSRIG LRVHFNLPED DKGSEASSE Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.

Striated muscle in vertebrates comprises large proteins which must be organized properly to contract efficiently. Z-lines in striated muscle are a sign of this organization, representing the ends of actin thin filaments, titin, nebulin or nebulette and accessory proteins required for structure and function. This gene encodes a protein which interacts with nebulin in skeletal muscle or nebulette in cardiac muscle and alpha-actinin. In addition, this gene product can interact with a protein with the I-band indicating it has a regulatory as well as structural function. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYPN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYPN
Gene Accession No. Q86TC9
Gene Alias 1110056A04Rik; 145 kDa sarcomeric protein; 9830112J03; AI853556; CMD1DD; CMH22; Kiaa4170; mKIAA4170; MYOP; Myopalladin; MYPN; RCM4; sarcomeric protein myopalladin, 145 kDa (MYOP); wu:fc17a02
Gene Symbols MYPN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYPN. Recombinant protein control fragment (Product #RP-103407).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84665
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.