Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYPN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116688 Supplier Invitrogen™ Supplier No. PA5116688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HepG2 Immunogen sequence: MQDDSIEAST SISQLLRESY LAETRHRGNN ERSRAEPSSN PCHFGSPSGA AEGGGGQDDL PDLSAFLSQE ELDESVNLAR LAINYDPLEK ADETQARKRL SPDQMKHSPN LSFEPNFCQD NPRSPTSSKE SPQEAKRPQY CSETQSKKVF LNKAADFIEE LSSLFKSHSS KRIRPRACKN HKSKLESQNK VMQENSSSFS DLSERRERSS VPIPIPADTR DNEVNHALEQ QEAKRREAEQ AASEAAGGDT.

Striated muscle in vertebrates comprises large proteins which must be organized properly to contract efficiently. Z-lines in striated muscle are a sign of this organization, representing the ends of actin thin filaments, titin, nebulin or nebulette and accessory proteins required for structure and function. This gene encodes a protein which interacts with nebulin in skeletal muscle or nebulette in cardiac muscle and alpha-actinin. In addition, this gene product can interact with a protein with the I-band indicating it has a regulatory as well as structural function. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYPN
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MYPN
Gene Accession No. Q5DTJ9, Q86TC9
Gene Alias 1110056A04Rik; 145 kDa sarcomeric protein; 9830112J03; AI853556; CMD1DD; CMH22; Kiaa4170; mKIAA4170; MYOP; Myopalladin; MYPN; RCM4; sarcomeric protein myopalladin, 145 kDa (MYOP); wu:fc17a02
Gene Symbols MYPN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human MYPN (NP_1159672).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68802, 84665
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.