Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYPOP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566761
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566761 Supplier Invitrogen™ Supplier No. PA566761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ACPQRYVLSED RREDRRADTS AHSKAGSSSP EPWARPSCTP QEGGCPRPKE RESPP Highest antigen sequence identity to the following orthologs - mouse 71%, rat 73%.

Transcriptional repressor; DNA-binding protein that specifically recognizes the core sequence 5'-YAAC[GT]G-3'. Dimerization with PFN1 reduces its DNA-binding capacity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYPOP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYPOP
Gene Accession No. Q86VE0
Gene Alias Myb related transcription factor, partner of profilin; myb-related protein p42POP; Myb-related transcription factor, partner of profilin; MYPOP; p42 Myb-related transcription factor, partner of profilin; P42pop; Partner of profilin
Gene Symbols MYPOP
Host Species Rabbit
Immunogen Recombinant Human MYPOP. Recombinant protein control fragment (Product #RP-107419).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339344
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.