Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYRIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544984 Supplier Invitrogen™ Supplier No. PA544984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QQRRKLPAPP VKAEKIETSS VTTIKTFNHN FILQGSSTNR TKERKGTTKD Sequence homology: Dog: 86%; Horse: 93%; Human: 100%; Pig: 93%.

MYRIP is a protein coding gene. Rab effector protein involved in melanosome transport. Serves as link between melanosome-bound RAB27A and the motor proteins MYO5A and MYO7A. May link RAB27A-containing vesicles to actin filaments. Functions as a protein kinase A-anchoring protein (AKAP). May act as a scaffolding protein that links PKA to components of the exocytosis machinery, thus facilitating exocytosis, including insulin release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYRIP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MYRIP
Gene Accession No. Q8NFW9
Gene Alias A230081N12Rik; DKFZp586F1018; exophilin; exophilin 8; exophilin-8; FLJ44025; MGC130034; MGC130035; myosin VIIA and Rab interacting protein; myosin-VIIa- and Rab-interacting protein; MYRIP; OTTHUMP00000209206; Rab effector MyRIP; Slac2c; slaC2-c; slp homolog lacking C2 domains c; Slp homologue lacking C2 domains; synaptotagmin-like protein homologue lacking C2 domains-c; synaptotagmin-like protein lacking C2 domains C
Gene Symbols MYRIP
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human MYRIP (aa 798-847).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25924
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.