Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYSM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578629
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578629 Supplier Invitrogen™ Supplier No. PA578629
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: HeLa, HepG2 Predicted Reactivity: Mouse (86%), Chicken (81%), Bovine (88%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Metalloprotease that specifically deubiquitinates monoubiquitinated histone H2A, a specific tag for epigenetic transcriptional repression, thereby acting as a coactivator. Preferentially deubiquitinates monoubiquitinated H2A in hyperacetylated nucleosomes. Deubiquitination of histone H2A leads to facilitate the phosphorylation and dissociation of histone H1 from the nucleosome. Acts as a coactivator by participating in the initiation and elongation steps of androgen receptor (AR)-induced gene activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYSM1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Mysm1
Gene Accession No. Q5VVJ2
Gene Alias 2ADUB; 2A-DUB; C130067A03Rik; C530050H10Rik; Histone H2A deubiquitinase MYSM1; KIAA1915; Myb like, SWIRM and MPN domains 1; myb-like, SWIRM and MPN domain-containing protein 1; myb-like, SWIRM and MPN domains 1; Mysm1; RGD1311787; RP4-592A1.1
Gene Symbols Mysm1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human MYSM1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114803
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.