Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYSM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA591929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA591929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA591929 Supplier Invitrogen™ Supplier No. PA591929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, A-431, HepG2, HeLa, Jurkat, Mouse brain, Rat brain; Cellular Location: Nucleus Immunogen sequence: SYRLPYKFEV QQMLEEPQWG LVFEKTRWII EKYRLSHSSV PMDKIFRRDS DLTCLQKLLE CMRKTLSKVT NCFMAEEFLT EIENLFLSNY KSNQENGVTE EN.

Metalloprotease that specifically deubiquitinates monoubiquitinated histone H2A, a specific tag for epigenetic transcriptional repression, thereby acting as a coactivator. Preferentially deubiquitinates monoubiquitinated H2A in hyperacetylated nucleosomes. Deubiquitination of histone H2A leads to facilitate the phosphorylation and dissociation of histone H1 from the nucleosome. Acts as a coactivator by participating in the initiation and elongation steps of androgen receptor (AR)-induced gene activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYSM1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Mysm1
Gene Accession No. Q5VVJ2, Q69Z66
Gene Alias 2ADUB; 2A-DUB; C130067A03Rik; C530050H10Rik; Histone H2A deubiquitinase MYSM1; KIAA1915; Myb like, SWIRM and MPN domains 1; myb-like, SWIRM and MPN domain-containing protein 1; myb-like, SWIRM and MPN domains 1; Mysm1; RGD1311787; RP4-592A1.1
Gene Symbols Mysm1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 720-821 of human MYSM1 (NP_0010789561).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114803, 298247, 320713
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.